missing translation for 'onlineSavingsMsg'
Learn More

Arsenic Polycrystalline lump, 2-8mm (0.08-0.3 in.), Puratronic™, 99.9999% (Metals basis), Thermo Scientific™

19.90 € - 3274.29 €

Chemical Identifiers

CAS 7440-38-2
Molecular Formula As
Molecular Weight (g/mol) 74.92
MDL Number MFCD00085309
InChI Key RQNWIZPPADIBDY-UHFFFAOYSA-N
Synonym arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
PubChem CID 5359596
ChEBI CHEBI:27563
IUPAC Name arsenic
SMILES [As]
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11336698
SDS View Documents
Thermo Scientific Alfa Aesar
013375.03
1 g
19.90 €
1g
Please to purchase this item. Need a web account? Register with us today!
11356698
SDS View Documents
Thermo Scientific Alfa Aesar
013375.09
10 g
103.52 €
10g
Please to purchase this item. Need a web account? Register with us today!
11376698
SDS View Documents
Thermo Scientific Alfa Aesar
013375.14
25 g
227.13 €
25g
Please to purchase this item. Need a web account? Register with us today!
11366698
SDS View Documents
Thermo Scientific Alfa Aesar
013375.22
100 g
640.19 €
100g
Please to purchase this item. Need a web account? Register with us today!
11346698
SDS View Documents
Thermo Scientific Alfa Aesar
013375.A1
1 kg
3274.29 €
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7440-38-2
74.92
RQNWIZPPADIBDY-UHFFFAOYSA-N
5359596
arsenic
As
MFCD00085309
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
CHEBI:27563
[As]

Specifications

7440-38-2
Polycrystalline Lump
As
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
[As]
74.92
CHEBI:27563
Gray lump
Gray
99.9999% (Metals basis)
MFCD00085309
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenic
5359596
Odorless
Arsenic
Videos
Safety and Handling

Safety and Handling

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: II

missing translation for 'einecsNumber' : 213-148-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.