missing translation for 'onlineSavingsMsg'
Learn More

Arsenic sponge, 30mm (1.18 in.) & down, 99.5% (metals basis), Thermo Scientific™

Product Code. 11324697
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-38-2
Molecular Formula As
Molecular Weight (g/mol) 74.92
MDL Number MFCD00085309
InChI Key RQNWIZPPADIBDY-UHFFFAOYSA-N
Synonym arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
PubChem CID 5359596
ChEBI CHEBI:27563
IUPAC Name arsenic
SMILES [As]

Specifications

UN Number UN1558
Quantity 25 g
Percent Purity 99.5%
Odor Odorless
Particle Size 30mm (1.18 in.) & down
Form Irregular Pieces
Assay (metals basis)
Chemical Name or Material Arsenic sponge
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'