Learn More
Arsenic standard solution, for AAS, 1mg/mL As in 2% KOH, Thermo Scientific Chemicals
48.14 € - 131.66 €
Specifications
Chemical Name or Material | Arsenic standard solution |
---|---|
Concentration | 997 to 1003 μg/mL of As(3+) in 2% KOH (20°C) |
Name Note | 1 mg/ml As in 2% KOH |
CAS | 7440-38-2, 1310-58-3, 7732-18-5 |
Packaging | Plastic bottle |
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
10452581
|
Thermo Scientific Acros
195821000 |
100 mL | Plastic bottle |
48.14 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10462581
|
Thermo Scientific Acros
195825000 |
500 mL | Plastic bottle |
131.66 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopySpezifikation
Arsenic standard solution | |
1 mg/ml As in 2% KOH | |
Plastic bottle | |
MFCD00085309 | |
RQNWIZPPADIBDY-UHFFFAOYSA-N | |
arsenic | |
5359596 | |
74.92 | |
1.02g/mL |
997 to 1003 μg/mL of As(3+) in 2% KOH (20°C) | |
7440-38-2, 1310-58-3, 7732-18-5 | |
As | |
15, 784 | |
[As] | |
74.92 | |
CHEBI:27563 | |
AAS | |
Solution |
Sicherheit und Handhabung
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Arsenic atomic absorption standard solution
Signalwort
- Danger
Gefahrenkategorie
- Skin corrosion/irritation Category 1 B
Gefahrenhinweis
- H314-Causes severe skin burns and eye damage.
Sicherheitshinweis
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.