Learn More
Thermo Scientific™ ART™ Barrier Low-Retention Pipette Tips
Product Code. 10314042
Click to view available options
Volume (Metric):
10 μL
20 μL
100 μL
200 μL
1000 μL
Model:
ART 10
ART 10 REACH
ART 1000
ART 100E
ART 10F
ART 200
ART 20P
Tip Style:
Ergonomic, Low Retention
Extended Length, Low Retention, Micro Point
Low Retention, Micro Point
Unit Size:
Pack of 800
Pack of 960
Fit for Purpose:
Sterile and PCR Ready
Product Code. 10314042
Brand: Thermo Scientific™ 2065E05
Only null left
Add to basket
Edge
Add to basket
Eliminate sample retention with these specially designed pipette tips.
Get Thermo Scientific™ ART™ Barrier Low-Retention Pipette Tips, which are specifically designed for eliminating sample retention within the pipette tip.
  • Inner surface is extremely hydrophobic, increasing sample reproducibility and sample accuracy
  • Crystal clear tips allow you to easily see transparent samples
  • Tips seal when exposed to potential contaminants
  • Certified RNase, DNase, ATP, Endotoxin, and Pyrogen Free
TRUSTED_SUSTAINABILITY
Specifications
Description ART 100E, Filtered, Sterile, Rack
Volume (Metric) 100 μL
Filtered Filtered
Sterility Sterile
Color Clear
Model ART 100E
Clean Claims DNase, RNase, ATP, Endotoxin and Pyrogen Free
Format Rack
Tip Style Low Retention, Micro Point
Packaging Rack, 10 x 96
Product Line ART