Learn More
Thermo Scientific™ ART™ Barrier Reload Insert Pipette Tips
GREENER_CHOICE
Obtain accurate and consistent performance with these pipette tips, which seal when exposed to potential contaminants.
Brand:  Thermo Scientific™ 2069-05-RI
77.22 EUR valid until 2025-05-31
Use promo code "25076" to get your promotional price.
 View more versions of this product
Product Code. 10308884
  • 122.00 € / Pack of 960
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Obtain accurate and consistent performance with Thermo Scientific™ ART™ Barrier Reload Insert Pipette Tips, which seal when exposed to potential contaminants. The ART inserts are environmentally friendly, providing individually packaged blister tubs that are made out of recycled plastic, designed to be used with the reusable ART hinged racks and creating the ART Reload System. The tips from the reload inserts clip securely into the hinged racks for efficient reloading.
Features
  • Sterile tips certified free from RNase, DNase and pyrogen
  • Available in MicroPoint, extended length, low retention and SoftFit-L™ designed for use with Rainin™ LTS™ LiteTouch™ Pipettes
Recommended for:
  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
TRUSTED_SUSTAINABILITY
Specifications
Specifications
200 μL
Sterile
2.32 in.
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Low Retention, Micro Point
Universal Fit
Sterile and PCR Ready
Filtered
ART 200
5.88cm
Refill
Fewer resources used and less waste
ART
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ ART™ Barrier Reload Insert Pipette Tips > ART Barrier Pipette tips 200; 200μL MicroPoint, reload insert, low retention, sterile tip