Learn More
Thermo Scientific™ ART™ Barrier Reload Insert, Extended Length Pipette Tips
GREENER_CHOICE
Obtain accurate and consistent performance with these pipette tips, which seal when exposed to potential contaminants.
Brand:  Thermo Scientific™ 2149E-05-RI
 View more versions of this product
Product Code. 10328444
  • 120.00 € / Pack of 960
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Obtain accurate and consistent performance with Thermo Scientific™ ART™ Barrier Reload Insert Pipette Tips, which seal when exposed to potential contaminants. The ART inserts are environmentally friendly, providing individually packaged blister tubs that are made out of recycled plastic, designed to be used with the reusable ART hinged racks and creating the ART Reload System. The tips from the reload inserts clip securely into the hinged racks for efficient reloading.
Features
  • Sterile tips certified free from RNase, DNase and pyrogen
  • Available in MicroPoint,extended length, low retention and SoftFit-L™ designed for use with Rainin™ LTS™ LiteTouch™ Pipettes
Recommended for:
  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ART 20E, Filtered, Sterile, Refill
Filtered
ART 20E
4.57cm
Refill
Fewer resources used and less waste
ART
10 μL
Sterile
1.8 in.
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Micro Point, Low Retention
Universal Fit Pipettor
Sterile & PCR Ready