Learn More
SCHOTT AG Lighting and Imaging™ Articulated Arm
This Articulated Arm is for use with base (158340) or directly on a microscope with M6-thread.
Brand:  SCHOTT AG Lighting and Imaging™ 158345
Product Code. 15723032
  • 272.00 € / 1 unit
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Articulated arm to position flexible light guides and lightlines precisely.
  • For flexible light guides.
  • Holders for different size light guides are available.
Specifications
Specifications
Articulated Arm
Base 158340 or Directly on a Microscope with M6-Thread.
7.9 in.
Stereo Microscopy Illumination
2 x M6 Holes
200 mm