missing translation for 'onlineSavingsMsg'
Learn More

Ascarite(II), CO2 absorbent, 8 to 20 mesh, Thermo Scientific Chemicals

127.64 € - 1301.00 €

Specifications

Color Brown
Physical Form Granular
Chemical Name or Material Ascarite(II)™, CO2 absorbent, 8 to 20 mesh
Name Note 8- to 20-mesh
CAS 81133-20-2, 1310-73-2, 7631-86-9
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10617504
View Documents
Thermo Scientific Acros
208081000
100 g
127.64 €
100g
Please to purchase this item. Need a web account? Register with us today!
10092680
View Documents
Thermo Scientific Acros
208085000
500 g
361.80 €
500g
Please to purchase this item. Need a web account? Register with us today!
15214684
View Documents
Thermo Scientific Acros
208080025
2.5 kg
1301.00 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Gas Adsorbent
Specifications

Specifications

Brown
Ascarite(II)™, CO2 absorbent, 8 to 20 mesh
81133-20-2, 1310-73-2, 7631-86-9
Particle size analysis: 8 to 20 mesh
Granular
8- to 20-mesh
MFCD00132264
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Product Identifier
  • Ascarite(II)Ò, CO2 absorbent, product of Arthur H. Thomas Company

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.

Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.