missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASGR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11739-1-AP-20UL
This item is not returnable.
View return policy
Description
The asialoglycooprotein receptor (ASGPR) is a transmembrane glycoprotein (42 kDa) which mediates binding, internalisation and degradation of extracellular glycoproteins that have exposed terminal galactose residues. The receptor is expressed on the surface of the hepatocytes in a polar manner, i.e. it is present on the sinusoidal and lateral palasma membranes but not on the bile canalicular membrane. The mammalian hepatic ASGPR mediate the endocytosis and degradation of serum proteins from which terminal sialic residues have been removed.The rat asialoglycoprotein receptor (ASGPR) consists of three polypeptide subunits: rat hepatic lectin (RHL)-1, RHL-2 and RHL-3. The monoclonal antibody 8D7 recognizes a subunit-specific epitope of RHL-1 of rat ASGPR. The monoclonal antibody 8D7 is cross reactive with human ASGPR.Specifications
ASGR1 | |
Polyclonal | |
Unconjugated | |
ASGR1 | |
ASGP R 1, ASGPR, ASGPR 1, Asgpr1, ASGR1, asialoglycoprotein receptor 1, CLEC4H1, Hepatic lectin H1, HL 1, Hs.12056 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11889, 24210, 432 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry, Immunoprecipitation | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P02706, P07306, P34927 | |
ASGR1 | |
ASGR1 Fusion Protein Ag2310 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |