Learn More
ASS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16210-1-AP-150UL
 View more versions of this product
Product Code. 16865754
  • 410.00 € / 150µL
Explore more special offers
Description
Description
The protein encoded by this gene catalyzes the penultimate step of the arginine biosynthetic pathway. There are approximately 10 to 14 copies of this gene including the pseudogenes scattered across the human genome, among which the one located on chromosome 9 appears to be the only functional gene for argininosuccinate synthetase. Mutations in the chromosome 9 copy of ASS cause citrullinemia. Two transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
ASS1
Polyclonal
Unconjugated
ASS1
Argininosuccinate synthase, argininosuccinate synthetase 1, ASS, ASS1, Citrulline aspartate ligase, CTLN1
Rabbit
Antigen Affinity Chromatography
RUO
11898, 25698, 445
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P00966, P09034, P16460
ASS1
ASS1 Fusion Protein Ag9200
150 μL
Primary
Human, Mouse, Rat