missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint-Gobain AstiPure™ RF1 Series PFA Male Tube Connector
Use these fittings to deliver corrosive fluids. Saint Gobain Performance Plastics™ AstiPure™ RF1 Series PFA Male Tube Connector are leak proof and reliable under a wide range of temperatures and pressures.
Brand: Saint-Gobain RF10613
This item is not returnable.
View return policy
Description
- Sturdy rubber Silicone piston rings for flasks
- For general lab applications

Specifications
PFA | |
For 6 x 8mm dia. tubing | |
13mm | |
Male Tube Connector |
46 mm | |
1 Each | |
1/4 in. NPT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Saint-Gobain AstiPure™ RF1 Series PFA Male Tube Connector > Length: 46mm; Thread Size: 1/4 in. NPT
Spot an opportunity for improvement?Share a Content Correction