missing translation for 'onlineSavingsMsg'
Learn More

ASTM (ASTM D6045, D1500) Colour Reference Standard, Paragon Scientific™

ASTM Colour Reference Standards are for routine calibration of colour measuring instruments and verification of test data. Process industries throughout the world use colorimeters for colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.

Brand:  Paragon Scientific 134030


 View more versions of this product

Product Code. 16070152

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Full Traceability to International Standards
  • 500mL Pack Size supplied with MSDS
  • 18 months shelf life
  • Tamper evident security packaging
  • Manufactured in the United kingdom
Specifications

Specifications

500 mL
Color Reference Standard
Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ASTM Colour Reference Standard

Signal Word
  • Danger

Hazard Category
  • Aspiration toxicity Category 1

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.

Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

Supplemental information
  • MIXTURE LIST-Contains : white mineral oil

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Nominal values quoted