Learn More
ASTM™ D6584 Monoolein solution, Honeywell Fluka™
5000 μg/mL in pyridine, analytical standard, reference material
Brand:  Honeywell Fluka R1381-3ML
Product Code. 15929701
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Specifications
110-86-1, 111-03-5
3 mL
0.98 g/cm3
20°C
Liquid
Glass Bottle
Analytical Standard
115°C
Biofuels