missing translation for 'onlineSavingsMsg'
Learn More

VEE GEE ASTM Specific Gravity Hydrometer

The ASTM Specific Gravity hydrometer is manufactured to the specifications of the American Society for Testing and Materials.

Brand:  VEE GEE 67112H

 View more versions of this product

Product Code. 17806493

  • 43.00 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Approx. Length: 330 mm
  • Calibrated for use at 15.5°C/60°F
Specifications

Specifications

Specific Gravity Hydrometer
0.0005
ASTM Specific Gravity Hydrometer, ASTM 112H, 1.050 to 1.100
1.050 to 1.100
112H
33 cm
Glass
15.5°C (60°F)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers