missing translation for 'onlineSavingsMsg'
Learn More

Atenolol, 98%, Thermo Scientific Chemicals

Product Code. 13575040
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29122-68-7
Molecular Formula C14H22N2O3
Molecular Weight (g/mol) 266.34
InChI Key METKIMKYRPQLGS-UHFFFAOYSA-N
Synonym atenolol, tenormin, tenormine, normiten, blokium, prenormine, tenoblock, atehexal, betablok, cuxanorm
PubChem CID 2249
ChEBI CHEBI:2904
IUPAC Name 2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetamide
SMILES CC(C)NCC(COC1=CC=C(C=C1)CC(=O)N)O

Specifications

Melting Point 146.0°C to 148.0°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Merck Index 15,85
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in methanol, acetic acid, dmso, ethanol, insoluble in acetonitrile, ethylacetate, chlorofor, m
Formula Weight 266.34
Percent Purity 98%
Chemical Name or Material Atenolol
Product Identifier
  • Atenolol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.