Learn More
uvex Athletic D5XP Safety Glove
Shop All Uvex Products
Click to view available options
Size:
6
7
8
9
10
11
Description
- Outer material made from HPPE, steel and polyamide/elastane
- Micro-NBR foam coating on palm and fingertips
- High cut protection (Level D)
- High mechanical resistance
- Good grip on dry and (slightly) oily/wet workpieces
- Flexible safety gloves with excellent fit
- Good tactile feel
- Safety glove with knitted cuff and reinforced thumb joint
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Certifications/Compliance | EN 388 (4 X 4 3 D) |
Coating Material | Nitrile Foam |
Color | Gray, Black |
Hand | Ambidextrous |
Latex Free | Yes |
Material | HPPE, Steel Fiber, Nylon, Elastane |
Product Type | Cut-resistant Gloves |
Size | 11 |
Product Content Correction
Product Title
uvex Athletic D5XP Safety Glove >
Spot an opportunity for improvement?Share a Content Correction