missing translation for 'onlineSavingsMsg'
Learn More

ATP disodium salt, Tocris Bioscience™

Product Code. 18093992
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

P2 agonist

P2 purinoceptor agonist.

Specifications

CAS 987-65-5
Molecular Formula C10H14N5Na2O13P3
Quantity 50 mg
Synonym 5'-atp,na4
InChI Key SXXXIPSHRYMXLS-IDIVVRGQSA-N
SMILES C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)[O-])[O-].[Na+].[Na+]
IUPAC Name disodium;(2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate
Molecular Weight (g/mol) 551.145
PubChem CID 131675547
Formula Weight 551.14
Percent Purity >99%
Chemical Name or Material ATP disodium salt
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'