Learn More
ATP5D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14893-1-AP-20UL
 View more versions of this product
Product Code. 16883784
  • 145.00 € / 20µL
Explore more special offers
Description
Description
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the delta subunit of the catalytic core. Alternatively spliced transcript variants encoding the same isoform have been identified.
Specifications
Specifications
ATP5D
Polyclonal
Unconjugated
ATP5D
ATP5D, F ATPase delta subunit
Rabbit
Antigen Affinity Chromatography
RUO
245965, 513, 66043
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P30049, P35434, Q9D3D9
ATP5D
ATP5D Fusion Protein Ag6682
20 μL
Primary
Mouse, Rat, Human