missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Atracurium besylate, 96%

Product Code. 15678735
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64228-81-5
Molecular Formula C53H72N2O12·2C6H5O3S
Molecular Weight (g/mol) 1243.49
InChI Key XXZSQOVSEBAPGS-UHFFFAOYSA-L
Synonym atracurium besylate, atracurium besilate, tracrium, atracurii besilas, besilate d'atracurium, besilato de atracurio, tracrium preservative free, atracurii besilas inn-latin, bw 33a
PubChem CID 47320
ChEBI CHEBI:2915
IUPAC Name benzenesulfonate;5-[3-[1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-2-methyl-3,4-dihydro-1H-isoquinolin-2-ium-2-yl]propanoyloxy]pentyl 3-[1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-2-methyl-3,4-dihydro-1H-isoquinolin-2-ium-2-yl]propanoate
SMILES C[N+]1(CCC2=CC(=C(C=C2C1CC3=CC(=C(C=C3)OC)OC)OC)OC)CCC(=O)OCCCCCOC(=O)CC[N+]4(CCC5=CC(=C(C=C5C4CC6=CC(=C(C=C6)OC)OC)OC)OC)C.C1=CC=C(C=C1)S(=O)(=O)[O-].C1=CC=C(C=C1)S(=O)(=O)[O-]

Specifications

Melting Point 90.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (mix of isomers) (HPLC)
Quantity 5 g
Formula Weight 1243.49
Percent Purity 96%
Chemical Name or Material Atracurium besylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'