missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 5 g |
Assay Percent Range | 95+% |
Merck Index | 15, 862 |
Melting Point | 175.0°C |
Solubility | Soluble in ether, chloroform and methanol, Very soluble in dimethyl sulfoxide, Insoluble in water |
Formula Weight | 215.69 |
Chemical Name or Material | Atrazine |
Safety and Handling



Product Identifier
- Atrazine
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H317-May cause an allergic skin reaction.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P314-Get medical advice/attention if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : Atrazine
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Atrazine, 95+%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction