Learn More
Heidolph Instruments Attachment for Test Tubes
For use with Reax top and Reax control
Brand:  Heidolph Instruments 549-01000-00
Product Code. 10049130
  • 46.15 € / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
For up to 10 test tubes with outer diameter of 10 mm and length up to 60 mm
Specifications
Specifications
Test Tubes
10 mm dia. x 60 mm L
Up to 10 test tubes with 10 mm diameter up to 60 mm length
10