missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Attachment for Bottles

For use with Reax 20/4, 20/8 and 20/12 shakers

Brand:  Heidolph™ 549-27000-00

 View more versions of this product

Product Code. 11721649

  • 92.00 € / Pack of 4
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Allows for the use of (4) 0.5 or 1 L bottles

Specifications

Specifications

0.5 L bottles
Reax 20/4, 20/8 and 20/12
4
4 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers