Learn More
Applied Biosystems™ Attune™ Flow Cytometer Syringe
The sample syringe is used with the fluidics system of the Attune Flow Cytometer to move sample to the flow cell.
Brand:  Applied Biosystems™ 4476090
 View more versions of this product
Product Code. 17246493
  • 1220.00 € / 10mL
Explore more special offers
Description
Description
  • The sample syringe is used with the fluidics system of the Attune Flow Cytometer to move sample to the flow cell.
  • Replacement of this syringe is recommended as part of instrument maintenance.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Attune™
10 mL
Attune Flow Cytometer
Store at room temperature
Room Temperature
Syringe