Learn More
Invitrogen™ Attune™ NxT Focus Bottle
This is a 1.9-L replacement bottle for the focusing fluid tank on the Attune NxT Acoustic Focusing Cytometer.
Brand: Invitrogen™ 100085929CST
This item is not returnable.
View return policy
Description
- This is a 1.9-L replacement bottle for the focusing fluid tank on the Attune NxT Acoustic Focusing Cytometer.
- The bottle is designed for easy detachment from the instrument for refill and with more robust fittings to prevent breakage.
- This bottle replaces discontinued Cat. No. 100022155.
Specifications
1 Bottle | |
Bottle | |
1 bottle | |
Flow Cytometer Accessory |
Attune NxT | |
FPL | |
Room temperature |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
Product Title
Invitrogen™ Attune™ NxT Focus Bottle > 1 bottle
Spot an opportunity for improvement?Share a Content Correction