Learn More
Attune™ NxT Maintenance Kit
The Attune NxT Maintenance Kit will help you avoid down-time and ensure optimal operation of your Attune NxT Acoustic Focusing Cytometer hardware.
Brand:  Invitrogen A43038
2213.50 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Product Code. 17174729
  • 2380.00 € / Each
Explore more special offers
Description
Description
This convenient kit includes sufficient supplies to perform all recommended preventative maintenance procedures for 12 months. These procedures are: replacement of the syringe (bi-yearly), SIP tube (yearly), and instrument bottles (yearly), as well as performance of system decontamination (quarterly). The kit includes 1 waste bottle, 1 focusing fluid tank, 12 focusing fluid filters, 2 syringes, 1 SIP tube, and 1 debubble solution.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Attune™
• 1 waste bottle
• 1 focusing fluid bottle
• 12 focusing fluid filters
• 2 syringes
• 1 SIP
• 1 debubble solution
Store at room temperature.
Approved for shipment at Room Temperature or on Wet Ice
Maintenance Kit
Non-Interchangeable
1 kit
Attune Acoustic Focusing Cytometer