Learn More
Attune™ NxT Red Laser Upgrade Kit
The Attune™ NxT Red Laser Upgrade Kit is designed to expand the 637 nm excitation capabilities of the Attune™ NxT Acoustic Focusing Cytometer.
Brand:  Invitrogen 100022778
Product Code. 15310667
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Given the large array of labels, dyes, and applications to choose from, the ability to upgrade your instrument is key. The Red Laser upgrade kit enables the detection of APC, Alexa Fluor™ 700, and APC-tandem dyes for immunophenotyping, as well as many other functional reagents such as CellTrace™ Far Red for cell proliferation, LIVE/DEAD™ Fixable Far Red or Near IR dyes for viability, Vybrant™ DyeCycle™ Ruby dye for live cell cycle analysis, and far red exciting fluorescent proteins.
Expanded capabilities
A great way to stretch your research dollars is through upgrades, thereby expanding the capabilities of your instrument while minimizing expenditure on new instruments. Through the upgrade path, the Attune™ NxT cytometer allows you to configure your instrument with different laser configurations to meet your ever-changing research needs.
Simple installation
The Attune™ NxT cytometer was designed to minimize the time it takes for a qualified service engineer to add additional capabilities. This Red Laser upgrade kit comes with all of the optical parts needed to upgrade the instrument in typically less than 4 hours.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Attune™
1 kit
Laser Upgrade Kit
Contains red laser with fiber optic cable and optics block.
Attune Acoustic Focusing Cytometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Attune™ NxT Red Laser Upgrade Kit > Contains red laser with fiber optic cable and optics block.