missing translation for 'onlineSavingsMsg'
Learn More

Attune™ Shutdown Solution (1X)

44.76 € - 135.00 €

Specifications

Content And Storage Store at room temperature.
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Shutdown Solution
Product Line Attune™
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15351086
View Documents
Invitrogen
A24975
250 mL
44.76 €
250mL
Please to purchase this item. Need a web account? Register with us today!
17334743
View Documents
Invitrogen
J106628
1 L
135.00 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Attune™ Shutdown Solution is a cleaning buffer that is used in the automated shutdown procedure for the Attune™ Acoustic Focusing Cytometer.

Specifications

Specifications

Store at room temperature.
Attune Acoustic Focusing Cytometer
Shutdown Solution
Attune™
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ATTUNE 1X SHUTDOWN SOLUTION

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 3(2H)-isothiazolone,5-chloro-2-methyl-with 2-methyl-3(2H)-isothiazolone

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.