Learn More
3M™ Aura™ Particulate Respirators 9300+ Series
Particulate respirators have an ergonomic, flat-fold, 3-panel design featuring a low breathing resistance filter media. Available in FFP1, FFP2, and FFP3 protection levels with or without valves, Innovative chin tab for easy adjustments and an embossed, sculpted upper panel helps reduce eyewear fogging.
Brand: 3M™ 7000062381
Description
Particulate respirators have an ergonomic, flat-fold, 3-panel design featuring a low breathing resistance filter media. Available in FFP1, FFP2, and FFP3 protection levels with or without valves, Innovative chin tab for easy adjustments and an embossed, sculpted upper panel helps reduce eyewear fogging.
- Provide comfortable protection against dusts and mists in a wide variety of industrial applications and other hazardous environments.
- Offers FFP1, FFP2 and FFP3 protection.
- 3-panel design allows for greater facial movement.
- Sculpted, low-profile style of the embossed upper panel creates an effective seal, reduces eyewear fogging and improves eyewear compatibility.
- Low resistance filter technology promotes easier breathing and the innovative chin tab allows simple adjustments.
- Choose without valve style or with valve styles that feature 3M™ Cool Flow™ to reduce heat build-up and help provide comfortable protection in hot and humid conditions.
- Individually packed in hygienic wrapping to prevent contamination before use.
- Braided, colour-coded headbands help you identify the protection level offered: yellow for FFP1 (APF 4), blue for FFP2 (APF 10) and red for FFP3 (APF 20).
- Foldable design makes them easy to store when not in use.
- Respirators work well with 3M eyewear and hearing protection products.

Specifications
4 | |
EN 149:2001 + A1:2009 | |
Flat-fold | |
FFP1 | |
General Purpose Particulate Respirator | |
Yes | |
Yes | |
PF9312 | |
Sculpted | |
Dusts and mists found in a wide variety of industrial applications and other work situations | |
3-panel | |
No |
EN 149:2001 + A1:2009 | |
White | |
Limited-use | |
Polypropylene | |
One Size | |
Polypropylene | |
Polyisoprene | |
Aluminum, Polyurethene | |
Individually Packaged | |
Non-sterile | |
Colour-coded Headband, Yellow |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.