missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Aura™ Particulate Respirator, 9300+Gen3 Series
Aura™ Particulate Respirator 9300+Gen3 Series is the next generation of 3M Aura Respirator: FFP protection combined with the original 3-panel, flat-fold design.
Brand: 3M™ 7100134298
This item is not returnable.
View return policy
Description
- Original 3-panel, flat-fold design fits a wide range of face shapes and sizes
- 3M Advanced Electret (filter) Media helps you breathe easier
- Innovative tabs on the upper and lower panels simplify positioning and adjustment
- Braided headbands are color-coded for quick and easy identification
- Sculpted upper panel conforms to the nose and contours of the face for an effective seal
- Embossed upper panel reduces eyewear fogging
- Easy to store away with flat-fold design and individual wrapping
- Breathing protection against hazardous work environments
- Made in the UK
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CE Approved | |
Flat Fold | |
No | |
No | |
Up to 12 x TLV | |
242 mm | |
212 mm |
White | |
Standard | |
FFP2 | |
9320+Gen3 | |
20/Pk. | |
87 mm | |
304 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction