Learn More
3M™ Aura™ Particulate Respirator, FFP3, Unvalved, 9330+
3M™ Aura™ Particulate Respirators 9300+ Series has an ergonomic, flat-fold, 3-panel design featuring a low breathing resistance filter media.
Brand:  3M™ 7000034741
Product Code. 30180450
  • 114.00 € / Pack of 20
Explore more special offers
Description
Description
  • Use 3M™ Aura™ Particulate Respirators 9300+ Series for comfortable protection against dusts and mists in a wide variety of industrial applications and other hazardous environments.
  • The 9300+ series offers FFP3 protection and allows you to talk and work in comfort.
  • The 3-panel design allows for greater facial movement, while the sculpted, low-profile style of the embossed upper panel creates an effective seal, reduces eyewear fogging and improves eyewear compatibility.
  • The low resistance filter technology helps you breathe easily and the innovative chin tab allows you to make simple adjustments.
  • Some of the range features a 3M™ Cool Flow™ Valve to reduce heat build-up and help provide comfortable protection in hot and humid conditions.
  • They're individually packed in hygienic wrapping to prevent contamination before use, and the braided, color-coded headbands help you identify the protection level offered: yellow for FFP1 (APF 4), blue for FFP2 (APF 10) and red for FFP3 (APF 20).
  • Their foldable design makes them easy to store when not in use, and they work well with 3M Eyewear and Hearing Protection products.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
White
Single-use
Composite Fabric
One Size
Flat-fold
FFP3
General Purpose Particulate Respirator
No