Learn More
3M™ Aura™ Particulate Respirator, Series 9400
Lightweight and foldable. The 3M Aura Particulate Respirator uses a unique three-panel design for greater comfort and easy communication.
Brand:  3M™ 7000088835
32.80 EUR valid until 2025-12-31
Use promo code "24304" to get your promotional price.
 View more versions of this product
Product Code. 12932505
  • 54.00 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Provide reliable and comfortable protection from a range of airborne disposable hazards
  • Minimize contamination risks to food or pharmaceutical production processes
  • Made without staples or small detachable parts
  • Cool Flow™ exhalation valve option provides effective removal of heat and moisture build up for a more comfortable wear
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EN 149:2001 + A1:2009
FFP2
FFP2
Cool Flow™ exhalation valve
Aluminum
Blue
Yes
Exposure to airborne duest particles and non-volatile liquid particles
Thermoplastic Elastomer
Airborne dust and non-volatile liquid particles