missing translation for 'onlineSavingsMsg'
Learn More

Slice™ Auto-Retractable Utility Scraper

Auto-retract scraper with SureScrape™ protects hand from excess strain. Durable finger-friendly™ safety blade is non-conductive, chemically inert, never rusts.

Brand:  Slice™ 10593

Product Code. 16463649

  • 49.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • SureScrape™ reduces hand strain
  • Installed blade type: #10526 rounded tip
  • Durable nylon handle with soft-touch comfort grips
  • Ambidextrous design
  • Finger-friendly™ edge lasts 11x longer than metal
  • Fewer blade changes = fewer injuries
  • Reduces injuries, lowers costs
  • Non-sparking, non-conductive, non-magnetic blade
  • Chemically inert blade never rusts
  • Oil- and lubricant-free blade
  • Compatible blades: 10526, 10528
  • Includes built-in lanyard hole and tool-free blade change
  • Ships with rounded-tip blade, ideal for scraping paint, rust, and adhesives
  • Specifications

    Specifications

    Scraper
    137 mm
    Black/Green
    1 ceramic rounded tip blade
    2.29 oz.
    2.88 in.
    5.39 in.
    GFN, TPEE, POM, Carbon Steel, Zirconium Oxide
    Safety Cutting
    1 Ea.
    64.8 g
    137 mm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.