missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific™ Autoclavable Bags

Clear, imprinted Autoclavable Bags with blue biohazard labeling specifically designed for safe disposal.
137.00 € - 264.00 €
Specifications
Product Type | Autoclavable Bags |
---|---|
Content And Storage | Room Temperature |
Description | Clear Autoclavable Bags |
Autoclavable | Autoclavable (121°C) |
Color | Clear |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
16370666
![]() |
Heathrow Scientific™
HS1002A |
310 x 660 mm |
137.00 €
Pack of 200 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
16380666
![]() |
Heathrow Scientific™
HS1002B |
415 x 600 mm |
143.00 €
Pack of 200 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
16390666
![]() |
Heathrow Scientific™
HS1002C |
610 x 810 mm |
264.00 €
Pack of 200 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
Description
- Constructed of 2.0 mm thick polypropylene.
- Can be used for repeated autoclaving.
- Strong side seams securely hold items.
- Autoclavable at 250°F (121°C).
- Front write-on panel.
- 4 inner packs of 50 bags.
- Disposable.
Specifications
Autoclavable Bags | |
Clear Autoclavable Bags | |
Clear | |
Hazardous Waste Disposal | |
Polypropylene | |
2 mm | |
50 Pk. | |
80 mil |
Room Temperature | |
Autoclavable (121°C) | |
Single-use | |
Autoclave | |
Non-sterile | |
200/Cs. | |
Box |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Heathrow Scientific™ Autoclavable Bags