missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Autoclavable Disposal Bags

Manufactured from Polypropylene or Polyamide. BRAND™ Autoclavable Disposal Bags are for collecting disposable items after use. Polypropylene bags are autoclavable at 121°C. Polyamide bags are autoclavable at 134°C or can be sterilized with hot air at 180°C

Brand:  BRAND™ 759705

14.45 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.


 View more versions of this product

Product Code. 10717091

  • 25.70 € / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Description

Description

For collection of disposable items after use. Caution! Risk of injury: Never put sharp objects such as needles into plastic bags.
  • Bags must be open during sterilization.
  • Polypropylene bags autoclavable at 121°C
  • Polyamide bags autoclavable at 134°C or can be sterilized with hot air at 180°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

300 mm
200 mm
Autoclaving items for disposal
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Autoclavable Disposal Bags > 100 Bags

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.