missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Polypropylene Straight Connector Barbed Fitting

These economical straight barbed fittings easily connect fluid paths in the laboratory without the use of tools.

Brand:  Masterflex™ 06365-22

 View more versions of this product

Product Code. 12459570

  • 56.90 € / Pack of 25
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Ideal for low-pressure applications, typically up to 30 psi (2 bar)
  • Chemical resistance
  • Slip flexible tubing or hose onto the barb to create a secure seal
  • No leaks, flow restrictions or costly downtime
  • Sterilization through atuoclaving or ethylene oxide (ETO)
Specifications

Specifications

Barbed Fitting
Polypropylene
25/Pk.
Autoclave or Ethylene Oxide
Straight connector
30 psi (2 bar)
Autoclavable
1/8 in. tubing I.D.
1/16 in. orifice; 27/32 x 13/32 in.
Straight
Clear
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.