missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Autoclavable Safe-Lock Micropestle
GREENER_CHOICE

Use this pestle for resuspending pellets. Eppendorf™ Autoclavable Safe-Lock Micropestles feature two sides for optimal fit in 1.5 and 2.0mL vessels.

Brand:  Eppendorf™ 0030120973

49.20 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10683001

  • 49.80 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use this pestle for resuspending pellets. Eppendorf™ Autoclavable Safe-Lock Micropestles feature two sides for optimal fit in 1.5 and 2.0mL vessels.

  • Autoclavable (121°C, 20 min.)
  • For use with 1.5 and 2.0mL vessels
  • Available in packs of ten
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Micropestle
For 1.5mL and 2mL Safe-Lock tubes and 1.7mL Safe-Twist screw cap tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf™ Autoclavable Safe-Lock Micropestle > For use with 1.5mL and 2mL Safe-Lock Tubes and 1.7mL Safe-Twist Screw Cap Tubes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.