Learn More
Microspec™ Autoclave Bags
Designed for contaminated waste disposal in autoclaves or incinerators.
Brand:  Microspec™ H1
 View more versions of this product
Product Code. 15722135
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Clear bags imprinted with blue biohazard labeling designed for safe disposal
  • Constructed of thick polypropylene
  • Allows for repeated autoclaving
  • Strong side seams for securely holding items
  • Front write-on panel
Specifications
Specifications
Autoclave Bags
Autoclavable
Single-use
Non-sterile
Polypropylene
EcoPak
Medium
632 mm
413 mm
62.733 lb.
Room Temperature
Clear with Blue Print
Polypropylene
50 μm
200
1500
2 mil
16.26 in.
413 mm
28.455 kg