Learn More
WLD Tec™ Autoloop Pro
Autoloop Pro - Fully automated carousel for flame sterilizing inoculation loops.
Brand:  WLD Tec™ 8.000.400
Product Code. 10744447
  • 1645.00 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The inoculation loop carousel Autoloop Pro, which is suitable for up to 4 inoculation loops, was developed to comply with current requirements in microbiological, cell-biological and biotechnological laboratories.
  • Modern Design: The stable housing enables comfortable and easy access to inoculation loops. Removal positions on both sides make the carousel equally suitable for right and left handers.
  • Saves Energy, Time and Costs: Precise flame sterilization time extends the life of the inoculation loops and reduces gas consumption. Continuous working during the flaming and the cooling phases of the inoculation loops saves up to 70% of time and makes it possible to work efficiently.
  • More Comfort: Keep all functions in view with the fully graphic display. The graphic menu control makes it easy to select all functions quickly. Flaming and cooling time can be adjusted to the second. During the flame sterilizing, the display shows the remaining flaming time. The Autoloop PRO rotates and controls flaming automatically. When flaming is completed, the display shows the remaining cooling time and the removal positions of cool inoculation loops. All the suspended inoculation loops can be flamed as needed in one full rotation of the carousel.
  • For additional flame sterilisation (e.g. of bottle necks or test tubes) a foot pedal can be activated during the automatic operation. For this purpose the Autoloop PRO turns the inoculation loop out of the flame position and enables additional flame sterilization.
  • More Safety: The nearly perpendicular position of the inoculation loop holder protects even the sleeve nut of the holder from possible contaminations. Additionally, an intelligent sensor of the Autoloop PRO monitors safe sterilization.
  • Robust: Fabricated entirely of stainless steel, anodized aluminum and a display, protected by heat resistant glass, the Autoloop PRO can withstand extreme laboratory conditions.
  • Less is more: The low gas consumption and the large savings in time set new standards for the laboratory.
Specifications
Specifications
Fuego SCS series
1200 g
Stainless Steel and Aluminum