Learn More
Applied Biosystems™ AutoLys M TubeLifter
The AutoLys M TubeLifter enables a rack of 24 AutoLys M Tubes to be simultaneously 'lifted' and readied for centrifugation.
Brand:  Applied Biosystems™ A37956
Product Code. 15900503
  • 3120.00 € / Each
Explore more special offers
Description
Description
Once centrifugation is complete the rack of tubes is returned to the TubeLifter and the inner tube of each AutoLys M Tube is separated from the outer tube. The outer tube contains the lysate that is processed further using the MagMAX FFPE DNA/RNA Ultra Kit to recover the nucleic acids.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AutoLys TubeLifter System
1 each
Room Temperature
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number5166257 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberA37956
Lot Number5170614 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberA37956
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ AutoLys M TubeLifter > AutoLys TubeLifter System