Learn More
ATAGO™ Automatic Digital Refractometer RX-007α
The Automatic Digital Refractometer RX-007α is suitable for measuring water soluble samples with very low concentrations, such as tea and low sugar beverages, at a very high accuracy.
Brand:  ATAGO™ 3921
Product Code. 10141123
  • 10715.00 € / Each
Estimated Shipment: 04-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The new RX-007α (alpha) is suitable for measuring water soluble samples with very low concentrations (5.000% or less) such as tea (green tea, black tea, oolong tea) and low sugar beverages at a very high accuracy of ±0.005%.
  • This product comes standard with a two (2) year limited warranty against manufacturer's defects from the date of the original purchase. This warranty does not cover or apply to the prism. The warranty period can be extended to three (3) years if the product is registered with ATAGO.
  • FDA 21 CFR Part 11 Software Included in Standard Delivery.
Specifications
Specifications
Benchtop Refractometer
±0.005% (Brix), 0.000010 (Refractive Index)
6.7 kg
0.001°C
1.330150 to 1.341500
0.001% (Brix), 0.000001 (Refractive Index)
370 x 260 x 140 mm
15.00°C to 30.00°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
ATAGO™ Automatic Digital Refractometer RX-007α > Refractometer; Refractive Index: 1.330150 to 1.341500