Learn More
Socorex™ Dosys™ Premium Pistol Grip Automatic Syringe
Dose with excellent reproducibility using a corrosion-resistant pistol-grip or two-ring handle automatic syringe. Socorex™ Dosys™ Premium Pistol Grip Automatic Syringe fits either hand comfortably and requires limited effort for efficient work even during long dispensing sessions.
Brand:  Socorex™ 174.0502
 View more versions of this product
Product Code. 10400021
  • 255.00 € / Each
Explore more special offers
Description
Description
  • Pistol grip handle.
  • Self-refilling automatic syringe.
  • Detachable Luer lock valve system.
  • High chemical resistance.
  • Corrosion resistant.
  • Optimal hand balance and ergonomics.
    • Fits either hand.
    • Requires limited effort.
  • Fast and reliable volume setting.
  • High reproducibility due to safety locking nut on a micrometric screw.
  • High-precision stainless steel plunger.
  • Ground glass barrel.
  • Smooth plunger travel and tightness without O-ring.
  • Easy disassembly for efficient cleaning and parts replacement if necessary.
  • Autoclavable (except for protective PVC sleeve).
Specifications
Specifications
Autoclavable
Syringe, 1M silicone feed tube, sinker, PVC protection sleeve, aspiration, vent and dosing cannulas, set of spares for valve, operating instructions.
0.1 mL
1/ea.
0.3 to 2 mL
Yes
Borosilicate Glass (Barrel), Stainless Steel (Plunger)
Automatic Syringe, Pistol Grip
Dosys™ Premium 174
<0.7% to <0.4%
Luer Lock