missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ Automation Pipette Tips for Biomek™ Instruments Workstations

These automation pipette tips come racked in different volume sizes for your convenience.

Brand:  Labcon™ 8024-690-010-9

 View more versions of this product

Product Code. 17587966

  • 595.00 € / Pack of 4800
Estimated Shipment: 12-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Pipette tips for automated workstations
  • Fits Beckman™ Biomek Workstations
  • Polypropylene USP Class VI Resin
  • Magenta color
  • Made in USA
Specifications

Specifications

90 μL
Non-sterile
Autoclavable
51.3 mm
ATP, RNase/Dnase, Endotoxin-free
4800 Tips
Automated Pipette System
APT
Non-filtered
Clear
2 in.
ISO 9001:2015, FDA CFR21 GMP, USP Class VI
Rack, 96
Single-use
No
Polypropylene Resin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers