Learn More
Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP384 Dispense Head
GREENER_CHOICE
Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Brand:  Thermo Scientific™ 935-262
4915.68 EUR valid until 2024-12-31
Use promo code "21898" to get your promotional price.
 View more versions of this product
Product Code. 11701273
  • 6005.00 € / Pack of 19200
Estimated Shipment: 08-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
  • Available in low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384- and 1536- deep well plates
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
30 μL
Non-sterile
Rack
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP384
Certified Quality
Non-filtered
Clear
Extended Length
Beckman