Learn More
Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
GREENER_CHOICE
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems through the superior tip straightness and low %CV.
Brand:  Thermo Scientific™ 2760
582.11 EUR valid until 2025-12-31
Use promo code "24288" to get your promotional price.
 View more versions of this product
Product Code. 11741353
  • 677.00 € / Pack of 4800
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well plates
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 μL
Sterile
Not Autoclavable
Free of RNase, DNase, DNA, ATP, and endotoxin
Extended Length
4800 tips
Beckman
Filtered
Clear
ISO 9001 and ISO 13485
Rack
96 tips/rack, 10 racks/pack, 5 packs/case
Universal Fit Pipettor
Sterile