missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for BioTek™ Pipetting Systems
GREENER_CHOICE

Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency.

430.92 € - 748.00 €

Specifications

Color Clear
Format Rack
For Use With (Equipment) BioTek Pipetting System
Product Line BioTek
View More Specs

Products 9
Product Code Brand Volume (Metric) Color Sterility Filtered Price Quantity & Availability  
Product Code Brand Volume (Metric) Color Sterility Filtered Price Quantity & Availability  
16456949
View Documents
Thermo Scientific™
984-021
50 μL Clear Sterile Filtered This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12358592
View Documents
Thermo Scientific™
984-261
50 μL Clear Sterile Non-filtered This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16466949
View Documents
Thermo Scientific™
984-262
50 μL Clear Non-sterile Non-filtered This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16476949
View Documents
Thermo Scientific™
985-021
80 μL Clear Sterile Filtered This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16486949
View Documents
Thermo Scientific™
985-261
100 μL Clear Sterile Non-filtered This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16496949
View Documents
Thermo Scientific™
985-262
100 μL Clear Non-sterile Non-filtered This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15243059
GREENER_CHOICE
View Documents
Thermo Scientific™
986-021
150 μL Clear Sterile Filtered
748.00 €
Pack of 4800
Estimated Shipment: 10-04-2025
to see stock.
Add to basket
 
12716027
View Documents
Thermo Scientific™
986-261
200 μL Clear Sterile Non-filtered This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16406959
View Documents
Thermo Scientific™
986-262
200 μL Clear Non-sterile Non-filtered This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: BioTek Precision and Precision XS.

Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Specifications

Specifications

Clear
Rack
BioTek Pipetting System
BioTek
Videos
SDS
Documents

Documents

Product Certifications