Learn More
Thermo Scientific™ Automation Tips for Qiagen™ Workstations
Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.
127.00 € - 642.00 €
Product Code | Brand | Volume (Metric) | Filtered | Packaging | Fit for Purpose | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Filtered | Packaging | Fit for Purpose | Price | Quantity & Availability | ||
10798302
|
Thermo Scientific™
951-011 |
250 μL | Filtered | 96 Tips/Tray, 10 Trays/Case | Sterile & PCR Ready |
213.00 €
Pack of 960 |
Estimated Shipment: 14-04-2025
Log in to see stock. |
||
10757922
|
Thermo Scientific™
951-021 |
250 μL | Filtered | 96 Tips/Tray, 10 Trays/Case | Sterile & PCR Ready |
172.00 €
Pack of 960 |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||
10063411
|
Thermo Scientific™
|
300 μL | Non-filtered | 96 Tips/Tray, 50 Trays/Case | Sterile |
642.00 €
Pack of 4800 |
Estimated Shipment: 07-03-2025
Log in to see stock. |
||
10274661
|
Thermo Scientific™
951-261 |
300 μL | Non-filtered | 96 Tips/Tray, 10 Trays/Case | Sterile |
136.00 €
Pack of 960 |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||
10081001
|
Thermo Scientific™
|
300 μL | Non-filtered | 96 Tips/Tray, 10 Trays/Case | Certified Quality |
127.00 €
Pack of 960 |
Estimated Shipment: 07-04-2025
Log in to see stock. |
||
10264711
|
Thermo Scientific™
953-021 |
1000 μL | Filtered | 96 Tips/Tray, 10 Trays/Case | Sterile & PCR Ready |
172.00 €
Pack of 960 |
Estimated Shipment: 07-03-2025
Log in to see stock. |
||
10071541
|
Thermo Scientific™
953-253 |
1100 μL | Non-filtered | 96 Tips/Tray, 50 Trays/Case | Sterile |
642.00 €
Pack of 4800 |
Estimated Shipment: 07-03-2025
Log in to see stock. |
||
10386691
|
Thermo Scientific™
953-261 |
1100 μL | Non-filtered | 96 Tips/Tray, 10 Trays/Case | Sterile |
136.00 €
Pack of 960 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||
10284661
|
Thermo Scientific™
953-262 |
1100 μL | Non-filtered | 96 Tips/Tray, 10 Trays/Case | Certified Quality |
153.00 €
Pack of 960 |
Estimated Shipment: 07-04-2025
Log in to see stock. |
||
Description
Increase pipetting performance with Thermo Scientific™ Automation Tips for Qiagen™ Universal Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Qiagen Rosys, Colibri and BioRobot 3000/8000/9600/9604.
Features:- Available as sterile and filtered.
- Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
- Lot-tested during production on actual workstation for which they are created.
- Low percentage coefficient of variation (%CV).
Warranty:: A five (5)-year shelf life from the date of manufacture is recommended.