Learn More
Trajan Scientific and Medical™ Autosampler Syringes with Cone Tipped Needle for Agilent GCs
Gives better performance and superior quality. Trajan Scientific and Medical Autosampler Syringes with Cone Tipped Needle for Agilent GCs has the knowledge and expertise to manufacture an SGE syringe.
Brand:  Trajan Scientific and Medical™ 002820
 View more versions of this product
Product Code. 10514365
  • 785.00 € / Pack of 10
Explore more special offers
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Agilent instruments
Specifications
Specifications
23 G
0.63 mm
10 μL
0.11 mm
Removable Needle Agilent Syringe with GT Plunger
Cone