Learn More
Trajan Scientific and Medical™ Autosampler Syringes with Cone Tipped Needle for Agilent GCs
Gives better performance and superior quality. Trajan Scientific and Medical Autosampler Syringes with Cone Tipped Needle for Agilent GCs has the knowledge and expertise to manufacture an SGE syringe.
Brand: Trajan Scientific and Medical™ 000610
This item is not returnable.
View return policy
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Agilent instruments
Specifications
23 G | |
0.63 mm | |
1 μL |
0.22 mm | |
NanoVolume Agilent Syringe | |
Cone |