Learn More
Trajan™ Autosampler Syringes with Removable Needle for CTC Analytics CombiPAL and GC PAL
For reduced non specific sample interaction and increased analyte recovery
Brand:  Trajan™ 004862
 View more versions of this product
Product Code. 13199205
  • 115.00 € / Each
Explore more special offers
Description
Description
  • Borosilicate glass barrel for strength and inertness.
  • Stainless steel needles for inertness, strength and corrosion resistance.
  • Backing behind the scale aids quick and accurate reading of the syringe scale.
  • Metal back flange provides stability and resists fracture.
  • Metal needle cap on syringe barrel protects and improves syringe lifetime.
  • Syringes are individually tested for quality ensuring reliability and conformance.
  • Unique packaging design eliminates possible contamination in the syringe.
Specifications
Specifications
23 G
0.63 mm
2.2 in.
50F-RTC/RSH-GT-5.7/0.63C
50 μL
Thermo RSH Autosampler
0.02 in.
Gas chromatography
Autosampler Syringe
Ea.
Cone
Fixed