missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Avidin, NeutrAvidin™, Alexa Fluor™ 350 conjugate
Avidin, NeutrAvidin™, Alexa Fluor™ 350 conjugate
Brand: Molecular Probes™ A11236
241.68 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
This item is not returnable.
View return policy
Description
Our Alexa Fluor 350 conjugate of NeutrAvidin biotin-binding protein – a form of avidin that has been processed to remove carbohydrate and to lower its isoelectric point – can substantially decrease background due to nonspecific binding. The method used to deglycosylate the avidin retains its specific binding. This bright, blue-fluorescent reagent (absorption/emission maxima ∼346/442 nm) is ideal for detecting biotinylated targets in fluorescence microscopy and flow cytometry applications.
Specifications
Alexa Fluor 350 | |
Store in freezer (-5 to -30°C) and protect from light. | |
Alexa Fluor™, NeutrAvidin™ |
1 mg | |
Room Temperature | |
Alexa Fluor Dyes |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Molecular Probes™ Avidin, NeutrAvidin™, Alexa Fluor™ 350 conjugate > 1mg
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction